This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03138269_g1
See other GSK3BA GE Assays ›
Gene Symbol:
gsk3ba
Gene Name:
glycogen synthase kinase 3 beta, genome duplicate a
Gene Aliases:
GSK-3[b], GSK3, fb68h05, fk80d11, gsk3b, wu:fb68h05, wu:fk80d11
Chromosome Location:
Chr.9: 9577743 - 9671325 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03138269_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: gsk3ba
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_131381.1 NP_571456.1 10 - 11 1890 68
XM_009304494.3 XP_009302769.1 10 - 11 1982 68
GenBank mRNA AB032265.1 - 10 - 11 1426 68
AJ223502.1 - 10 - 11 1890 68
BC162371.1 - 10 - 11 1823 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
heart looping
heart jogging
protein phosphorylation
signal transduction
insulin receptor signaling pathway
regulation of neuron projection development
phosphorylation
regulation of microtubule cytoskeleton organization
negative regulation of canonical Wnt signaling pathway
extrinsic apoptotic signaling pathway in absence of ligand
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
ATP binding
beta-catenin binding
kinase activity
transferase activity
tau-protein kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0