This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03138267_mH
See other GSK3BA GE Assays ›
Gene Symbol:
gsk3ba
Gene Name:
glycogen synthase kinase 3 beta, genome duplicate a
Gene Aliases:
GSK-3[b], GSK3, fb68h05, fk80d11, gsk3b, wu:fb68h05, wu:fk80d11
Chromosome Location:
Chr.9: 9577743 - 9671325 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03138267_mH
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: gsk3ba
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_131381.1 NP_571456.1 8 - 9 1609 108
GenBank mRNA AB032265.1 - 8 - 9 1145 108
AJ223502.1 - 8 - 9 1609 108
BC162371.1 - 8 - 9 1542 108

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
heart looping
heart jogging
protein phosphorylation
signal transduction
insulin receptor signaling pathway
regulation of neuron projection development
phosphorylation
regulation of microtubule cytoskeleton organization
negative regulation of canonical Wnt signaling pathway
extrinsic apoptotic signaling pathway in absence of ligand
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
ATP binding
beta-catenin binding
kinase activity
transferase activity
tau-protein kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header