This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03138177_m1
See other SEMA3H GE Assays ›
Gene Symbol:
sema3h
Gene Name:
sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3H
Gene Aliases:
Sema3G, sema8, semaZ8
Chromosome Location:
Chr.8: 25913432 - 25958448 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03138177_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: sema3h
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001328378.1 NP_001315307.1 8 - 9 981 90
NM_131050.2 NP_571125.2 8 - 9 981 90
XM_009303983.3 XP_009302258.1 8 - 9 1336 90
GenBank mRNA AY860954.1 - 8 - 9 903 90
BC162689.1 - 8 - 9 981 90
CU638804.1 - 8 - 9 903 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

membrane-bound signaling molecule intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

neural crest cell migration
axon guidance
motor neuron axon guidance
positive regulation of cell migration
negative regulation of axon extension involved in axon guidance
negative chemotaxis
semaphorin-plexin signaling pathway
positive regulation of neuron migration
semaphorin receptor binding
neuropilin binding
chemorepellent activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0