This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03136182_m1
See other KCNH1A GE Assays ›
Gene Symbol:
kcnh1a
Gene Name:
potassium voltage-gated channel, subfamily H (eag-related), member 1a
Gene Aliases:
Eag1, Kv10.1, kcnh1
Chromosome Location:
Chr.17: 25677346 - 25770231 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03136182_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: kcnh1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001044931.1 NP_001038396.1 10 - 11 2028 64
XM_009293096.3 XP_009291371.1 12 - 13 2234 64
XM_009293097.2 XP_009291372.1 12 - 13 2234 64
XM_009293098.3 XP_009291373.1 11 - 12 2207 64
XM_009293099.3 XP_009291374.1 11 - 12 2207 64
GenBank mRNA BC162812.1 - 10 - 11 2068 64
DQ022612.1 - 10 - 11 2028 64
HQ703597.1 - 10 - 11 2028 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

ion channel

Gene Ontology Categories:

Function(s) Process(es)

somitogenesis
ion transport
potassium ion transport
hindbrain formation
regulation of ion transmembrane transport
regulation of membrane potential
chordate embryonic development
transmembrane transport
potassium ion transmembrane transport
ion channel activity
voltage-gated ion channel activity
voltage-gated potassium channel activity
potassium channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0