This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03136117_m1
See other STIM1A GE Assays ›
Gene Symbol:
stim1a
Gene Name:
stromal interaction molecule 1a
Gene Aliases:
si:dkey-24p1.5
Chromosome Location:
Chr.15: 38221146 - 38297737 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03136117_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: stim1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001044799.1 NP_001038264.1 12 - 13 1789 79
XM_009291871.3 XP_009290146.1 13 - 14 1920 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

store-operated calcium entry
cellular calcium ion homeostasis
activation of store-operated calcium channel activity
axon extension involved in axon guidance
regulation of calcium ion transport
calcium channel regulator activity
calcium ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0