This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03135185_g1
See other SEMA3GB GE Assays ›
Gene Symbol:
sema3gb
Gene Name:
sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3Gb
Gene Aliases:
-
Chromosome Location:
Chr.8: 23485205 - 23554124 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03135185_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: sema3gb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001014824.1 NP_001014824.1 7 - 8 795 86
XM_005167032.4 XP_005167089.1 8 - 9 1408 86
XM_005167033.4 XP_005167090.1 8 - 9 1408 86
GenBank mRNA AY766121.1 - 7 - 8 795 86
BC163245.1 - 7 - 8 827 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

neural crest cell migration
axon guidance
positive regulation of cell migration
negative regulation of axon extension involved in axon guidance
negative chemotaxis
semaphorin-plexin signaling pathway
semaphorin receptor binding
chemorepellent activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0