This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03132335_g1
See other RPS6KAL GE Assays ›
Gene Symbol:
rps6kal
Gene Name:
ribosomal protein S6 kinase a, like
Gene Aliases:
rps6ka6, wu:fj94b02, zgc:66139
Chromosome Location:
Chr.14: 8947450 - 9002405 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03132335_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: rps6kal
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_200073.1 NP_956367.1 7 - 8 698 116
XM_009307718.2 XP_009305993.1 2 - 3 195 116
GenBank mRNA BC045478.1 - 7 - 8 803 116
BC057467.1 - 7 - 8 698 116

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
phosphorylation
intracellular signal transduction
nucleotide binding
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
ribosomal protein S6 kinase activity
ATP binding
kinase activity
transferase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0