This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03131833_g1
See other ATP6V1E1B GE Assays ›
Gene Symbol:
atp6v1e1b
Gene Name:
ATPase H+ transporting V1 subunit E1b
Gene Aliases:
atp6e, atp6v1e1, chunp6932, fj35f08, wu:fj35f08
Chromosome Location:
Chr.4: 5240126 - 5247266 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03131833_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: atp6v1e1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_173254.1 NP_775361.1 2 - 3 290 116
GenBank mRNA AF506222.1 - 2 - 3 290 116
BC057254.1 - 2 - 3 292 116
BC067557.1 - 2 - 3 299 116

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

fin regeneration
caudal fin development
notochord cell vacuolation
retina development in camera-type eye
hydrogen ion transmembrane transport
hydrogen-exporting ATPase activity, phosphorylative mechanism
hydrogen:potassium-exchanging ATPase activity
proton-transporting ATPase activity, rotational mechanism

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0