This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03131671_m1
See other UBE2IA GE Assays ›
Gene Symbol:
ube2ia
Gene Name:
ubiquitin-conjugating enzyme E2Ia
Gene Aliases:
Ubc9-2, ube2i2, ube2ib
Chromosome Location:
Chr.3: 45365066 - 45373515 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03131671_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ube2ia
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_131833.2 NP_571908.1 5 - 6 487 87
XM_021469272.1 XP_021324947.1 4 - 5 424 87
GenBank mRNA AF332623.1 - 4 - 5 329 87
BC058302.1 - 5 - 6 487 87
BC066609.1 - 5 - 6 465 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

heart looping
cell cycle
chromosome segregation
multicellular organism development
protein sumoylation
embryonic heart tube elongation
cell division
definitive hemopoiesis
nucleotide binding
ATP binding
transferase activity
SUMO conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0