This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03129771_g1
See other SPPL2 GE Assays ›
Gene Symbol:
sppl2
Gene Name:
signal peptide peptidase-like 2
Gene Aliases:
SPPL2B, wu:fc16e01, wu:fc85d12, zgc:136525
Chromosome Location:
Chr.2: 37480654 - 37503010 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03129771_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: sppl2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001045244.2 NP_001038709.2 10 - 11 1179 87
XM_021478906.1 XP_021334581.1 10 - 11 1174 87
XM_021478911.1 XP_021334586.1 10 - 11 1174 87
GenBank mRNA AY926457.1 - 10 - 11 1163 87
BC044512.1 - 10 - 11 1147 87
BC116574.1 - 10 - 11 1164 87
EH571140.1 - 4 - 5 483 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

aspartic protease

Gene Ontology Categories:

Function(s) Process(es)

membrane protein intracellular domain proteolysis
membrane protein proteolysis
negative regulation of coagulation
aspartic-type endopeptidase activity
aspartic endopeptidase activity, intramembrane cleaving

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0