This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03129761_m1
See other CRB2B GE Assays ›
Gene Symbol:
crb2b
Gene Name:
crumbs cell polarity complex component 2b
Gene Aliases:
crb2l
Chromosome Location:
Chr.8: 2826599 - 2910247 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03129761_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: crb2b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001045162.1 NP_001038627.1 9 - 10 3095 108
XM_017357360.2 XP_017212849.1 9 - 10 3137 108
GenBank mRNA DQ314737.1 - 9 - 10 3095 108
JQ740885.1 - 3 - 4 1727 108

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

cell adhesion molecule

Gene Ontology Categories:

Function(s) Process(es)

glomerular filtration
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
glomerulus development
slit diaphragm assembly
eye photoreceptor cell development
establishment or maintenance of epithelial cell apical/basal polarity
photoreceptor cell maintenance
pronephros development
retina morphogenesis in camera-type eye
cilium morphogenesis
glomerular visceral epithelial cell development
glomerular capillary formation
cell-cell adhesion
regulation of nonmotile primary cilium assembly
calcium ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0