This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03129623_m1
See other CACNA2D1A GE Assays ›
Gene Symbol:
cacna2d1a
Gene Name:
calcium channel, voltage-dependent, alpha 2/delta subunit 1a
Gene Aliases:
si:ch211-251k4.1
Chromosome Location:
Chr.4: 19764095 - 19884157 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03129623_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: cacna2d1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001044960.2 NP_001038425.2 24 - 25 2129 80
XM_005164746.4 XP_005164803.1 24 - 25 2305 80
XM_009300285.3 XP_009298560.1 25 - 26 2331 80
XM_009300286.3 XP_009298561.1 25 - 26 2328 80
XM_009300287.3 XP_009298562.1 25 - 26 2329 80
XM_009300288.3 XP_009298563.1 24 - 25 2307 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

voltage-gated ion channel

Gene Ontology Categories:

Process(es)

voltage-gated calcium channel activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0