This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03129518_m1
See other CHD4A GE Assays ›
Gene Symbol:
chd4a
Gene Name:
chromodomain helicase DNA binding protein 4a
Gene Aliases:
im:7143343, si:ch211-51m24.3
Chromosome Location:
Chr.19: 5000885 - 5058908 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03129518_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: chd4a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001044858.1 NP_001038323.1 25 - 26 3999 99
XM_005159352.3 XP_005159409.1 25 - 26 3996 99
XM_021468085.1 XP_021323760.1 25 - 26 3999 99

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

fin regeneration
RNA polymerase II repressing transcription factor binding
ATP binding
histone deacetylase binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0