This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03127419_g1
See other ZDHHC3A GE Assays ›
Gene Symbol:
zdhhc3a
Gene Name:
zinc finger DHHC-type palmitoyltransferase 3a
Gene Aliases:
ik:tdsubc_2d6, tdsubc_2d6, xx:tdsubc_2d6, zdhhc3, zgc:92587
Chromosome Location:
Chr.16: 39196694 - 39233451 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03127419_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: zdhhc3a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001002725.1 NP_001002725.1 2 - 3 456 68
XM_005158209.4 XP_005158266.1 2 - 3 628 68
GenBank mRNA BC076087.1 - 2 - 3 456 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein targeting to membrane
peptidyl-L-cysteine S-palmitoylation
palmitoyltransferase activity
transferase activity
transferase activity, transferring acyl groups
protein-cysteine S-myristoyltransferase activity
protein-cysteine S-palmitoyltransferase activity
protein homodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0