This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03127235_m1
See other FGF6A GE Assays ›
Gene Symbol:
fgf6a
Gene Name:
fibroblast growth factor 6a
Gene Aliases:
fgf4, fgf6
Chromosome Location:
Chr.25: 16945348 - 16951890 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03127235_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: fgf6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001001398.2 NP_001001398.2 2 - 3 855 63
GenBank mRNA AF516334.1 - 2 - 3 442 63
DQ317445.1 - 2 - 3 855 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

growth factor intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein phosphorylation
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
organ morphogenesis
positive regulation of gene expression
cell differentiation
regulation of cell migration
fibroblast growth factor receptor binding
growth factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0