This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03127194_m1
See other ASIC4B GE Assays ›
Gene Symbol:
asic4b
Gene Name:
acid-sensing (proton-gated) ion channel family member 4b
Gene Aliases:
ASIC4, ASIC4-B, accn4b, zASIC4.2, zgc:111839
Chromosome Location:
Chr.6: 13606417 - 13688949 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03127194_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: asic4b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_214786.1 NP_999951.1 8 - 9 1709 93
GenBank mRNA AJ609620.1 - 8 - 9 1709 93
BC095568.1 - 8 - 9 1716 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

ion channel

Gene Ontology Categories:

Function(s) Process(es)

ion transport
sodium ion transport
sodium ion transmembrane transport
response to steroid hormone
cation channel activity
sodium channel activity
ligand-gated sodium channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0