This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03127060_m1
See other PRPF4BB GE Assays ›
Gene Symbol:
prpf4bb
Gene Name:
pre-mRNA processing factor 4Bb
Gene Aliases:
prpf4b, wu:fc18e10, wu:fe47g02, zgc:55623
Chromosome Location:
Chr.24: 11473568 - 11505969 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03127060_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: prpf4bb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_213449.1 NP_998614.1 10 - 11 2341 92
GenBank mRNA BC048879.1 - 10 - 11 2341 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
protein kinase activity
ATP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0