This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03126774_m1
See other TIA1 GE Assays ›
Gene Symbol:
tia1
Gene Name:
TIA1 cytotoxic granule-associated RNA binding protein
Gene Aliases:
fb73d10, wu:fb73d10, wu:fc02b04, zgc:55520
Chromosome Location:
Chr.5: 305757 - 310746 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03126774_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: tia1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_212628.1 NP_997793.1 5 - 6 551 60
XM_009301867.2 XP_009300142.1 5 - 6 610 60
XM_009301868.3 XP_009300143.1 5 - 6 589 60
GenBank mRNA BC045368.1 - 5 - 6 573 60
BC152184.1 - 5 - 6 541 60

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_009301868.3 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Process(es)

nucleic acid binding
RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0