This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03126552_m1
See other IMPDH2 GE Assays ›
Gene Symbol:
impdh2
Gene Name:
IMP (inosine 5'-monophosphate) dehydrogenase 2
Gene Aliases:
IMPD 2, IMPDH 2, cb635, wu:fb64g02, wu:fc43f09
Chromosome Location:
Chr.8: 26059563 - 26079140 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03126552_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: impdh2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_201464.1 NP_958872.1 10 - 11 1310 76
XM_005166988.4 XP_005167045.1 11 - 12 1481 76
XM_021478183.1 XP_021333858.1 12 - 13 1555 76
XM_021478184.1 XP_021333859.1 11 - 12 1493 76
GenBank mRNA BC046905.1 - 10 - 11 1310 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

purine nucleotide biosynthetic process
GMP biosynthetic process
circadian rhythm
oxidation-reduction process
nucleotide binding
catalytic activity
IMP dehydrogenase activity
oxidoreductase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0