This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03125337_m1
See other NEO1A GE Assays ›
Gene Symbol:
neo1a
Gene Name:
neogenin 1a
Gene Aliases:
cb333, neo1, zgc:109795, zneo1
Chromosome Location:
Chr.7: 53754652 - 54089325 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03125337_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: neo1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001328497.1 NP_001315426.1 28 - 29 4542 85
NM_173218.2 NP_775325.2 28 - 29 4599 85
XM_005166588.3 XP_005166645.1 28 - 29 4608 85
XM_009303464.3 XP_009301739.1 28 - 29 4665 85
XM_009303465.3 XP_009301740.1 28 - 29 4663 85
XM_009303466.3 XP_009301741.1 27 - 28 4632 85
XM_009303468.3 XP_009301743.1 26 - 27 4414 85
XM_017356809.2 XP_017212298.1 27 - 28 4507 85
GenBank mRNA AF394058.1 - 28 - 29 85
AY029280.1 - 28 - 29 4547 85
AY082380.1 - 28 - 29 4489 85

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

somitogenesis
neural tube formation
nervous system development
axon guidance
positive regulation of BMP signaling pathway
iron ion homeostasis
signaling receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0