This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03123517_g1
See other AHSA1A GE Assays ›
Gene Symbol:
ahsa1a
Gene Name:
AHA1, activator of heat shock protein ATPase homolog 1a
Gene Aliases:
ahsa1, wu:fb83b05, zgc:136821, zgc:56075
Chromosome Location:
Chr.20: 26966927 - 26974181 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03123517_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ahsa1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001045091.1 NP_001038556.1 8 - 9 1111 132
XM_005160576.4 XP_005160633.1 8 - 9 1411 132
GenBank mRNA BC114279.1 - 8 - 9 1119 132

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein folding
positive regulation of ATPase activity
embryonic neurocranium morphogenesis
ATPase activator activity
chaperone binding
Hsp90 protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0