This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03123440_m1
See other RIPOR2 GE Assays ›
Gene Symbol:
ripor2
Gene Name:
RHO family interacting cell polarization regulator 2
Gene Aliases:
fam65b, si:dkey-218n20.1
Chromosome Location:
Chr.19: 31593386 - 31740953 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03123440_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ripor2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001045026.1 NP_001038491.1 7 - 8 741 114
XM_005159634.4 XP_005159691.1 7 - 8 788 114
XM_009294297.3 XP_009292572.1 7 - 8 655 114
XM_021468090.1 XP_021323765.1 7 - 8 903 114
XM_021468091.1 XP_021323766.1 7 - 8 811 114

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

chemotaxis
cell adhesion
negative regulation of cell adhesion
muscle organ development
sensory perception of sound
negative regulation of signal transduction
cell differentiation
negative regulation of Rho protein signal transduction
positive regulation of myoblast differentiation
skeletal muscle fiber development
positive regulation of filopodium assembly
positive regulation of neutrophil chemotaxis
positive regulation of myoblast fusion
negative regulation of establishment of T cell polarity
negative regulation of protein localization to cell leading edge
cellular response to chemokine
regulation of establishment of cell polarity
positive regulation of neutrophil extravasation
negative regulation of T cell migration
negative regulation of Rho guanyl-nucleotide exchange factor activity
14-3-3 protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0