This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03122671_g1
See other APOBB.1 GE Assays ›
Gene Symbol:
apobb.1
Gene Name:
apolipoprotein Bb, tandem duplicate 1
Gene Aliases:
apobb, apobl, cb235, hm:zeh1207, sb:cb235, si:ch211-219i10.1, wu:fb39c11, wu:fc51e12, zeh1207
Chromosome Location:
Chr.20: 31269702 - 31284894 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03122671_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: apobb.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001030062.1 NP_001025233.1 17 - 18 2870 60
XM_005160629.3 XP_005160686.1 18 - 19 2973 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

triglyceride mobilization
lipid transport
cholesterol transport
cholesterol homeostasis
lipoprotein transport
cellular response to xenobiotic stimulus
lipid transporter activity
low-density lipoprotein particle receptor binding
intermembrane cholesterol transfer activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0