This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03121972_g1
See other HDHD2 GE Assays ›
Gene Symbol:
hdhd2
Gene Name:
haloacid dehalogenase-like hydrolase domain containing 2
Gene Aliases:
zgc:110332
Chromosome Location:
Chr.10: 14529804 - 14540560 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03121972_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: hdhd2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001013473.1 NP_001013491.1 4 - 5 447 81
XM_017357962.2 XP_017213451.1 4 - 5 459 81
XM_017357963.2 XP_017213452.1 3 - 4 428 81
GenBank mRNA BC091457.1 - 4 - 5 447 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Function(s) Process(es)

dephosphorylation
phosphatase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0