This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03120924_mH
See other ACTR2A GE Assays ›
Gene Symbol:
actr2a
Gene Name:
actin related protein 2a
Gene Aliases:
actr2, hm:zeh1257, zgc:63719
Chromosome Location:
Chr.1: 51407471 - 51428111 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03120924_mH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: actr2a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_213499.1 NP_998664.1 7 - 8 927 58
XM_021470433.1 XP_021326108.1 8 - 9 991 58
GenBank mRNA BC055220.1 - 7 - 8 927 58

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

actin filament organization
Arp2/3 complex-mediated actin nucleation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of double-strand break repair via homologous recombination
regulation of double-strand break repair via nonhomologous end joining
nucleotide binding
actin binding
ATP binding
actin filament binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0