This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03118453_g1
See other SMYHC2 GE Assays ›
Gene Symbol:
smyhc2 LOC110438495
Gene Name:
slow myosin heavy chain 2
myosin-6-like
Gene Aliases:
MYH7, zgc:171631
-
Chromosome Location:
Chr.24: 40680153 - 40700596 on Build GRCz11
Chromosome Location:
Chr.Unknown: 4680 - 9287 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
-
Assay ID Dr03118453_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: smyhc2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001102626.2 NP_001096096.2 3 - 4 279 114
GenBank mRNA BC150362.1 - 3 - 4 279 114
Gene Symbol: LOC110438495
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021471019.1 XP_021326694.1 3 - 4 353 114

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

slow-twitch skeletal muscle fiber contraction
nucleotide binding
motor activity
actin binding
ATP binding
actin filament binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0