This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03118056_m1
See other BUD23 GE Assays ›
Gene Symbol:
bud23
Gene Name:
BUD23 rRNA methyltransferase and ribosome maturation factor
Gene Aliases:
wbscr22, zgc:162306
Chromosome Location:
Chr.15: 1749843 - 1765116 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03118056_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: bud23
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001082879.1 NP_001076348.1 9 - 10 744 65
GenBank mRNA BC133941.1 - 9 - 10 744 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

methyltransferase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

rRNA (guanine-N7)-methylation
methyltransferase activity
rRNA (guanine) methyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0