This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03117769_m1
See other NFIA GE Assays ›
Gene Symbol:
nfia
Gene Name:
nuclear factor I/A
Gene Aliases:
si:ch211-88d2.2, wu:fq27e07, zgc:158351
Chromosome Location:
Chr.22: 16835752 - 17053602 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03117769_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: nfia
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001079962.1 NP_001073431.1 2 - 3 786 60
XM_009296236.3 XP_009294511.1 2 - 3 1868 60
XM_009296237.3 XP_009294512.1 2 - 3 1868 60
XM_009296239.3 XP_009294514.1 2 - 3 1868 60
XM_009296240.3 XP_009294515.1 2 - 3 1868 60
XM_009296241.3 XP_009294516.1 2 - 3 1482 60
XM_009296242.3 XP_009294517.1 2 - 3 1868 60
XM_009296243.3 XP_009294518.1 2 - 3 1868 60
GenBank mRNA BC128847.1 - 2 - 3 786 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
DNA replication
regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0