This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03116921_m1
See other DVL1B GE Assays ›
Gene Symbol:
dvl1b
Gene Name:
dishevelled segment polarity protein 1b
Gene Aliases:
dvl1, dvl2l
Chromosome Location:
Chr.11: 24360582 - 24428628 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03116921_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: dvl1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001044917.1 NP_001038382.1 8 - 9 896 86
XM_005169297.4 XP_005169354.1 8 - 9 1314 86
XM_005169298.4 XP_005169355.1 8 - 9 1314 86
XM_005169299.4 XP_005169356.1 6 - 7 1132 86
XM_005169301.4 XP_005169358.1 8 - 9 1314 86
GenBank mRNA BC171461.1 - 8 - 9 1005 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

Wnt signaling pathway
intracellular signal transduction
canonical Wnt signaling pathway
planar cell polarity pathway involved in neural tube closure
frizzled binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0