This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03116437_m1
See other TPRB GE Assays ›
Gene Symbol:
tprb
Gene Name:
translocated promoter region b, nuclear basket protein
Gene Aliases:
si:dkey-97o5.5, tpr, wu:fi76g05
Chromosome Location:
Chr.20: 34039281 - 34069959 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03116437_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: tprb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001030123.1 NP_001025294.1 9 - 10 968 82
XM_009294751.3 XP_009293026.1 9 - 10 989 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

primary active transporter

Gene Ontology Categories:

Function(s) Process(es)

mRNA export from nucleus
protein import into nucleus
regulation of mitotic spindle assembly
structural constituent of nuclear pore

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0