Assay Details
Gene Symbol: tprb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030123.1 | NP_001025294.1 | 34 - 35 | 4996 | 107 | |
XM_009294751.3 | XP_009293026.1 | 34 - 35 | 5017 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
