This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03114208_g1
See other EWSR1B GE Assays ›
Gene Symbol:
ewsr1b LOC110438494 LOC110440168 clpxb
Gene Name:
EWS RNA-binding protein 1b
RNA-binding protein EWS-like
RNA-binding protein FUS-like
caseinolytic mitochondrial matrix peptidase chaperone subunit Xb
Gene Aliases:
fb40b11, fusl, wu:fb40b11, wu:fb75g09, zgc:55864
-
-
fc31b05, fj85g10, wu:fc31b05, wu:fj85g10
Chromosome Location:
Chr.5: 24873560 - 24882357 on Build GRCz11
Chromosome Location:
Chr.Unknown: 5098 - 7426 on Build GRCz11
Chromosome Location:
Chr.13: 135271 - 179227 on Build GRCz11
Chromosome Location:
Chr.25: 35574757 - 35599792 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
-
-
-
Assay ID Dr03114208_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ewsr1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_212630.1 NP_997795.1 11 - 12 1179 75
XM_005165107.2 XP_005165164.2 11 - 12 1166 75
XM_005165108.2 XP_005165165.2 11 - 12 1138 75
XM_009301286.1 XP_009299561.1 10 - 11 1134 75
XM_009301287.1 XP_009299562.1 10 - 11 1128 75
GenBank mRNA BC044518.1 - 11 - 12 1179 75
BC067661.1 - 10 - 11 1133 75
BC097019.1 - 11 - 12 1172 75
Gene Symbol: LOC110438494
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021471017.1 XP_021326692.1 4 - 5 445 75
Gene Symbol: LOC110440168
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021480686.1 XP_021336361.1 14 - 15 1567 75
XM_021480687.1 XP_021336362.1 14 - 15 1561 75
XM_021480688.1 XP_021336363.1 14 - 15 1540 75
XM_021480689.1 XP_021336364.1 13 - 14 1408 75
XM_021480691.1 XP_021336366.1 13 - 14 1446 75
XM_021480692.1 XP_021336367.1 13 - 14 1342 75
XM_021480693.1 XP_021336368.1 13 - 14 1315 75
XM_021480694.1 XP_021336369.1 11 - 12 1144 75
XM_021480695.1 XP_021336370.1 11 - 12 1138 75
XM_021480696.1 XP_021336371.1 8 - 9 733 75
Gene Symbol: clpxb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AI545751.1 - 2 - 3 233 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

mitotic cell cycle
regulation of transcription, DNA-templated
central nervous system neuron development
nucleic acid binding
transcription cofactor activity
RNA binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0