This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03113987_m1
See other CACNA1DA GE Assays ›
Gene Symbol:
cacna1da
Gene Name:
calcium channel, voltage-dependent, L type, alpha 1D subunit, a
Gene Aliases:
cacna1d, cav1.3a, isl, si:dz136d19.3, si:dz223c6.1
Chromosome Location:
Chr.11: 36792831 - 36964176 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03113987_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: cacna1da
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_203484.1 NP_982351.1 22 - 23 2864 89
XM_021479576.1 XP_021335251.1 22 - 23 3052 89
XM_021479578.1 XP_021335253.1 22 - 23 3052 89
XM_021479579.1 XP_021335254.1 22 - 23 3052 89
XM_021479581.1 XP_021335256.1 21 - 22 2976 89
XM_021479583.1 XP_021335258.1 22 - 23 3052 89
XM_021479584.1 XP_021335259.1 21 - 22 2976 89
GenBank mRNA AY528224.1 - 22 - 23 2864 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

voltage-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

ion transport
calcium ion transport
regulation of ion transmembrane transport
synapse organization
neuromuscular process controlling balance
detection of mechanical stimulus involved in sensory perception of sound
detection of mechanical stimulus involved in sensory perception
transmembrane transport
calcium ion import
calcium ion transmembrane transport
ion channel activity
voltage-gated ion channel activity
voltage-gated calcium channel activity
calcium channel activity
high voltage-gated calcium channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0