This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03111806_mH
See other ACVR2AA GE Assays ›
Gene Symbol:
acvr2aa
Gene Name:
activin A receptor type 2Aa
Gene Aliases:
actr2a, acvr2, acvr2a, wu:fa02e10
Chromosome Location:
Chr.9: 25380790 - 25444359 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03111806_mH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: acvr2aa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001110278.2 NP_001103748.2 9 - 10 2215 108
XM_005167561.4 XP_005167618.1 9 - 10 2525 108
XM_021478661.1 XP_021334336.1 10 - 11 2555 108
XM_021478662.1 XP_021334337.1 10 - 11 2506 108
GenBank mRNA AY962561.1 - 8 - 9 1161 108
D38262.1 - 9 - 10 2209 108

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

serine/threonine protein kinase receptor

Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
neural crest cell migration
protein phosphorylation
transmembrane receptor protein serine/threonine kinase signaling pathway
transforming growth factor beta receptor signaling pathway
phosphorylation
hindbrain development
activin receptor signaling pathway
cartilage development
cellular response to growth factor stimulus
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
transmembrane receptor protein serine/threonine kinase activity
ATP binding
kinase activity
transferase activity
activin-activated receptor activity
signaling receptor activity
metal ion binding
activin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0