This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03110548_m1
See other PCSK5A GE Assays ›
Gene Symbol:
pcsk5a
Gene Name:
proprotein convertase subtilisin/kexin type 5a
Gene Aliases:
pc5.1, pcsk5.1, pcsk5/6a, si:dkey-267i17.1
Chromosome Location:
Chr.5: 127333 - 215542 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03110548_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: pcsk5a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021476634.1 XP_021332309.1 21 - 22 2962 61
GenBank mRNA DQ983232.1 - 21 - 22 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
protein processing
peptide hormone processing
neuromast development
anterior lateral line development
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity
hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0