This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03110110_g1
See other SLC25A23B GE Assays ›
Gene Symbol:
slc25a23b
Gene Name:
solute carrier family 25 member 23b
Gene Aliases:
si:dkey-204f11.59
Chromosome Location:
Chr.3: 34220194 - 34231082 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03110110_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: slc25a23b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001044952.1 NP_001038417.1 9 - 10 1269 72
XM_021470903.1 XP_021326578.1 6 - 7 836 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

mitochondrial carrier protein

Gene Ontology Categories:

Function(s) Process(es)

transmembrane transport
ATP transmembrane transporter activity
calcium ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0