This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03110029_g1
See other SETDB1A GE Assays ›
Gene Symbol:
setdb1a
Gene Name:
SET domain bifurcated histone lysine methyltransferase 1a
Gene Aliases:
SETDB1, fc07d03, si:ch211-81a5.6, wu:fc07d03
Chromosome Location:
Chr.19: 9111548 - 9144392 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03110029_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: setdb1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001044767.2 NP_001038232.1 11 - 12 2948 117

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

chromatin organization
Ras protein signal transduction
negative regulation of gene expression
methylation
histone lysine methylation
histone H3-K9 methylation
heterochromatin organization
positive regulation of methylation-dependent chromatin silencing
DNA binding
methyltransferase activity
zinc ion binding
transferase activity
histone-lysine N-methyltransferase activity
metal ion binding
histone methyltransferase activity (H3-K9 specific)
promoter-specific chromatin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0