This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03110003_m1
See other PTPN4B GE Assays ›
Gene Symbol:
ptpn4b
Gene Name:
protein tyrosine phosphatase non-receptor type 4b
Gene Aliases:
fi40g11, ptpn4a, si:rp71-1n18.2, wu:fi40g11
Chromosome Location:
Chr.22: 11779834 - 11847366 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03110003_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ptpn4b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001044313.2 NP_001037778.2 15 - 16 1529 119
XM_005173946.4 XP_005174003.1 16 - 17 1925 119
XM_009296096.3 XP_009294371.1 16 - 17 1922 119

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
dephosphorylation
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
cytoskeletal protein binding
hydrolase activity
phosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0