This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03107811_m1
See other NRP2B GE Assays ›
Gene Symbol:
nrp2b
Gene Name:
neuropilin 2b
Gene Aliases:
id:ibd2759, npn-2B, sb:cb513
Chromosome Location:
Chr.9: 14367578 - 14504971 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03107811_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: nrp2b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_212966.1 NP_998131.1 14 - 15 2953 68
XM_017357663.2 XP_017213152.1 14 - 15 3082 68
GenBank mRNA AY351913.1 - 14 - 15 2450 68
AY428802.1 - 14 - 15 2953 68
AY437404.1 - 14 - 15 2913 68
BC163576.1 - 14 - 15 2768 68
BC163589.1 - 14 - 15 2768 68
KM655620.1 - 14 - 15 2363 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
vasculogenesis
neural crest cell migration
multicellular organism development
nervous system development
axon guidance
axonal fasciculation
cell differentiation
regulation of vascular endothelial growth factor receptor signaling pathway
vascular endothelial growth factor signaling pathway
olfactory bulb axon guidance
vascular endothelial growth factor-activated receptor activity
semaphorin receptor activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0