This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03107620_m1
See other SLC9A3R1A GE Assays ›
Gene Symbol:
slc9a3r1a
Gene Name:
SLC9A3 regulator 1a
Gene Aliases:
cb895, nherf1, slc9a3, slc9a3r1, slc9a3r2, wu:fd20a02, zgc:55541, zgc:77629
Chromosome Location:
Chr.12: 46747 - 107864 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03107620_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: slc9a3r1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_212707.2 NP_997872.2 1 - 2 606 82
GenBank mRNA BC045377.1 - 1 - 2 592 82
BC064290.1 - 1 - 2 584 82
BC164812.1 - 1 - 2 488 82
EH434174.1 - 1 - 2 635 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein targeting to membrane
ventricular system development
receptor clustering
sodium ion homeostasis
otic vesicle development
protein localization to plasma membrane
receptor binding
protein anchor
binding, bridging

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0