This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03107502_m1
See other AHSA1B GE Assays ›
Gene Symbol:
ahsa1b
Gene Name:
AHA1, activator of heat shock protein ATPase homolog 1b
Gene Aliases:
ahsa1l, id:ibd5076, wu:fb38h08
Chromosome Location:
Chr.17: 43850516 - 43863700 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03107502_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ahsa1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_212602.1 NP_997767.1 3 - 4 519 149
GenBank mRNA BC055523.1 - 3 - 4 519 149
BC068225.1 - 3 - 4 149
BC164374.1 - 3 - 4 392 149

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein folding
positive regulation of ATPase activity
ATPase activator activity
chaperone binding
Hsp90 protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0