This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03107329_m1
See other UCHL1 GE Assays ›
Gene Symbol:
uchl1
Gene Name:
ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase)
Gene Aliases:
cb358, wu:fc55h08
Chromosome Location:
Chr.1: 550931 - 551387 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03107329_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: uchl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_201477.1 NP_958885.1 4 - 5 371 72
GenBank mRNA AY147851.1 - 4 - 5 374 72
BC049044.1 - 4 - 5 371 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

cysteine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
ubiquitin-dependent protein catabolic process
protein deubiquitination
thiol-dependent ubiquitin-specific protease activity
peptidase activity
cysteine-type peptidase activity
hydrolase activity
ligase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0