This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03107268_m1
See other YARS1 GE Assays ›
Gene Symbol:
yars1
Gene Name:
tyrosyl-tRNA synthetase 1
Gene Aliases:
cb204, wu:fk52b08, yars
Chromosome Location:
Chr.19: 30884955 - 30895274 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03107268_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: yars1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_201316.1 NP_958473.1 6 - 7 656 79
XM_005159598.4 XP_005159655.1 6 - 7 681 79
XM_021468004.1 XP_021323679.1 6 - 7 647 79
GenBank mRNA AY398430.1 - 6 - 7 656 79
BC076558.1 - 6 - 7 635 79
BC154288.1 - 5 - 6 923 79
BC164691.1 - 5 - 6 626 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

translational protein

Gene Ontology Categories:

Function(s) Process(es)

translation
tRNA aminoacylation for protein translation
tyrosyl-tRNA aminoacylation
cellular response to DNA damage stimulus
tRNA binding
nucleotide binding
RNA binding
aminoacyl-tRNA ligase activity
tyrosine-tRNA ligase activity
ATP binding
ligase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0