This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03105195_m1
See other ALG12 GE Assays ›
Gene Symbol:
alg12
Gene Name:
ALG12 alpha-1,6-mannosyltransferase
Gene Aliases:
si:dkey-164m14.3
Chromosome Location:
Chr.4: 61546 - 70443 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03105195_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: alg12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001098749.1 NP_001092219.1 1 - 2 179 112
GenBank mRNA BC142932.1 - 1 - 2 179 112

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein N-linked glycosylation
dolichol-linked oligosaccharide biosynthetic process
alpha-1,6-mannosyltransferase activity
mannosyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
dolichyl-pyrophosphate Man7GlcNAc2 alpha-1,6-mannosyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0