This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03104381_m1
See other MDH1B GE Assays ›
Gene Symbol:
mdh1b
Gene Name:
malate dehydrogenase 1B, NAD (soluble)
Gene Aliases:
zgc:153922
Chromosome Location:
Chr.1: 6218628 - 6225363 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03104381_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: mdh1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001076761.1 NP_001070229.1 8 - 9 1181 91
XM_009305644.3 XP_009303919.1 7 - 8 1332 91
XM_009305648.3 XP_009303923.1 5 - 6 976 91
XM_021475844.1 XP_021331519.1 7 - 8 1278 91
GenBank mRNA BC124491.1 - 8 - 9 1181 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
tricarboxylic acid cycle
oxaloacetate metabolic process
malate metabolic process
NADH metabolic process
oxidation-reduction process
catalytic activity
oxidoreductase activity
malate dehydrogenase activity
oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
L-malate dehydrogenase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0