This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03103443_m1
See other SID4 GE Assays ›
Gene Symbol:
sid4
Gene Name:
secreted immunoglobulin domain 4
Gene Aliases:
-
Chromosome Location:
-
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03103443_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: sid4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001034182.2 NP_001029354.2 1115 79
GenBank mRNA AY494978.1 - 875 79
BC134848.1 - 1115 79
CU458937.1 - 899 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Inventoried

Gene Ontology Categories:

Function(s) Process(es)

anatomical structure morphogenesis
protein binding
identical protein binding
extracellular matrix binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0