This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03102985_mH
See other DNMT3BB.3 GE Assays ›
Gene Symbol:
dnmt3bb.3
Gene Name:
DNA (cytosine-5-)-methyltransferase beta, duplicate b.3
Gene Aliases:
dnmt3-2, dnmt5, wu:fc08a10, zgc:136325
Chromosome Location:
Chr.23: 16961269 - 16981980 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03102985_mH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: dnmt3bb.3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001020479.1 NP_001018315.1 15 - 16 3025 80
XM_005162039.4 XP_005162096.1 15 - 16 3078 80
XM_021469750.1 XP_021325425.1 14 - 15 2913 80
GenBank mRNA AB196916.1 - 15 - 16 3025 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

DNA methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

methylation
methyltransferase activity
transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0