This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03102338_m1
See other PPP2R1BB GE Assays ›
Gene Symbol:
ppp2r1bb
Gene Name:
protein phosphatase 2, regulatory subunit A, beta b
Gene Aliases:
ik:tdsubc_2h3, ppp2r1b, tdsubc_2h3, wu:fa11a11, wu:fb38a08, xx:tdsubc_2h3, zgc:92493
Chromosome Location:
Chr.15: 21744743 - 21772837 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03102338_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ppp2r1bb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001005590.1 NP_001005590.1 5 - 6 653 102
GenBank mRNA BC081658.1 - 5 - 6 653 102
BC097118.1 - 5 - 6 698 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons

Gene Ontology Categories:

Process(es)

protein serine/threonine phosphatase activity
protein phosphatase regulator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0