This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03102223_m1
See other SLC6A22.1 GE Assays ›
Gene Symbol:
slc6a22.1
Gene Name:
solute carrier family 6 member 22, tandem duplicate 1
Gene Aliases:
si:ch211-263a23.7, si:zc263a23.7, slc6a13
Chromosome Location:
Chr.6: 41118903 - 41138937 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03102223_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: slc6a22.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001004533.1 NP_001004533.1 11 - 12 1386 86
XM_005166019.4 XP_005166076.1 13 - 14 1745 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

primary active transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
sodium ion transmembrane transport
neurotransmitter:sodium symporter activity
gamma-aminobutyric acid:sodium symporter activity
symporter activity
neurotransmitter binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0