This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03101916_g1
See other NAXE GE Assays ›
Gene Symbol:
naxe
Gene Name:
NAD(P)HX epimerase
Gene Aliases:
apoa1bp, zgc:92263
Chromosome Location:
Chr.19: 7408275 - 7412647 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03101916_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: naxe
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001002618.1 NP_001002618.1 3 - 4 453 74
GenBank mRNA BC075969.1 - 3 - 4 453 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Gene Ontology Categories:

Process(es)

nucleotide binding
protein binding
isomerase activity
metal ion binding
NADHX epimerase activity
NADPHX epimerase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0